Assay Details
| Gene Symbol: ifnphi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207640.1 | NP_997523.1 | 2 - 3 | 240 | 104 | |
| GenBank mRNA | AB128865.1 | - | 2 - 3 | 104 | ||
| AJ544822.1 | - | 2 - 3 | 210 | 104 | ||
| AY135716.1 | - | 2 - 3 | 210 | 104 | ||
| BC162493.1 | - | 2 - 3 | 223 | 104 | ||
| EF014960.1 | - | 3 - 4 | 194 | 104 | ||
More Information
Set Membership: |
|

ifnphi1
Genomic Map