Assay Details
| Gene Symbol: cdk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212564.2 | NP_997729.1 | 6 - 7 | 777 | 72 | |
| GenBank mRNA | AF268044.1 | - | 6 - 7 | 804 | 72 | |
| BC079527.1 | - | 6 - 7 | 777 | 72 | ||
More Information
Set Membership: |
|

cdk1
Genomic Map