Assay Details
| Gene Symbol: bhlhe40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212679.2 | NP_997844.2 | 4 - 5 | 616 | 81 | |
| GenBank mRNA | AB212759.1 | - | 4 - 5 | 479 | 81 | |
| AY652763.1 | - | 4 - 5 | 616 | 81 | ||
| BC066738.1 | - | 4 - 5 | 594 | 81 | ||
More Information
Set Membership: |
|

bhlhe40
Genomic Map