Assay Details
| Gene Symbol: mecp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212736.2 | NP_997901.1 | 1 - 2 | 46 | 63 | |
| GenBank mRNA | AY298900.2 | - | 1 - 2 | 32 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mecp2| Gene Symbol: mecp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212736.2 | NP_997901.1 | 1 - 2 | 46 | 63 | |
| GenBank mRNA | AY298900.2 | - | 1 - 2 | 32 | 63 | |
Set Membership: |
|