Assay Details
| Gene Symbol: mecp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328555.1 | NP_001315484.1 | 2 - 3 | 418 | 76 | |
| NM_212736.2 | NP_997901.1 | 2 - 3 | 421 | 76 | ||
| GenBank mRNA | AY298900.2 | - | 2 - 3 | 407 | 76 | |
| BC093116.1 | - | 2 - 3 | 418 | 76 | ||
More Information
Set Membership: |
|

mecp2
Genomic Map