Assay Details
| Gene Symbol: fahd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212918.1 | NP_998083.1 | 6 - 7 | 929 | 94 | |
| XM_021478226.1 | XP_021333901.1 | 6 - 7 | 841 | 94 | ||
| XM_021478227.1 | XP_021333902.1 | 6 - 7 | 990 | 94 | ||
| XM_021478228.1 | XP_021333903.1 | 6 - 7 | 841 | 94 | ||
| XM_021478229.1 | XP_021333904.1 | 6 - 7 | 925 | 94 | ||
| GenBank mRNA | BC067576.1 | - | 6 - 7 | 929 | 94 | |
More Information
Set Membership: |
|

fahd2a
Genomic Map