Assay Details
| Gene Symbol: caprin1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213068.1 | NP_998233.1 | 8 - 9 | 1012 | 71 | |
| XM_021467626.1 | XP_021323301.1 | 8 - 9 | 1029 | 71 | ||
| XM_021467627.1 | XP_021323302.1 | 8 - 9 | 1032 | 71 | ||
| GenBank mRNA | BC047196.1 | - | 8 - 9 | 1012 | 71 | |
More Information
Set Membership: |
|

caprin1b
Genomic Map