Assay Details
| Gene Symbol: pck2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213192.1 | NP_998357.1 | 7 - 8 | 1288 | 63 | |
| GenBank mRNA | BC063985.1 | - | 7 - 8 | 1288 | 63 | |
More Information
Set Membership: |
|

pck2
Genomic Map