Assay Details
| Gene Symbol: hnrnpabb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213302.1 | NP_998467.1 | 4 - 5 | 513 | 87 | |
| GenBank mRNA | BC066681.1 | - | 4 - 5 | 513 | 87 | |
More Information
Set Membership: |
|

hnrnpabb
Genomic Map