Assay Details
| Gene Symbol: smpd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213355.1 | NP_998520.1 | 10 - 11 | 974 | 87 | |
| GenBank mRNA | BC057525.1 | - | 10 - 11 | 974 | 87 | |
More Information
Set Membership: |
|

smpd4
Genomic Map