Assay Details
| Gene Symbol: srsf6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213466.1 | NP_998631.1 | 4 - 5 | 541 | 71 | |
| XM_005161922.4 | XP_005161979.1 | 4 - 5 | 71 | |||
| XM_021469776.1 | XP_021325451.1 | 3 - 4 | 71 | |||
| GenBank mRNA | BC055238.1 | - | 4 - 5 | 541 | 71 | |
More Information
Set Membership: |
|

srsf6a
Genomic Map