Assay Details
| Gene Symbol: fgf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214808.1 | NP_999973.1 | 2 - 3 | 260 | 115 | |
| GenBank mRNA | AY457142.1 | - | 2 - 3 | 260 | 115 | |
| BC083269.1 | - | 2 - 3 | 135 | 115 | ||
| BC165029.1 | - | 2 - 3 | 105 | 115 | ||
More Information
Set Membership: |
|

fgf17
Genomic Map