Assay Details
| Gene Symbol: cenpn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002341.1 | NP_001002341.1 | 9 - 10 | 981 | 69 | |
| GenBank mRNA | AY648831.1 | - | 9 - 10 | 980 | 69 | |
| BC075923.1 | - | 9 - 10 | 981 | 69 | ||
More Information
Set Membership: |
|

cenpn
Genomic Map