Assay Details
| Gene Symbol: mrps26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002418.1 | NP_001002418.1 | 3 - 4 | 616 | 124 | |
| GenBank mRNA | BC076154.1 | - | 3 - 4 | 616 | 124 | |
More Information
Set Membership: |
|

mrps26
Genomic Map