Assay Details
| Gene Symbol: prkab1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002632.1 | NP_001002632.1 | 4 - 5 | 516 | 79 | |
| XM_005155628.4 | XP_005155685.1 | 4 - 5 | 583 | 79 | ||
| GenBank mRNA | BC075947.1 | - | 4 - 5 | 516 | 79 | |
More Information
Set Membership: |
|

prkab1a
Genomic Map