Assay Details
| Gene Symbol: znf106a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003200378.4 | XP_003200426.2 | 3 - 4 | 292 | 88 | |
| GenBank mRNA | BC076401.1 | - | 3 - 4 | 290 | 88 | |
| CB363924.1 | - | 3 - 4 | 290 | 88 | ||
| EB924828.1 | - | 3 - 4 | 238 | 88 | ||
| EB935832.1 | - | 3 - 4 | 277 | 88 | ||
More Information
Set Membership: |
|

znf106a
Genomic Map