Assay Details
| Gene Symbol: znf106a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003200378.4 | XP_003200426.2 | 4 - 5 | 486 | 111 | |
| GenBank mRNA | BC076401.1 | - | 4 - 5 | 111 | ||
| CB363924.1 | - | 4 - 5 | 111 | |||
| EB894275.1 | - | 1 - 2 | 186 | 111 | ||
| EB935832.1 | - | 4 - 5 | 471 | 111 | ||
More Information
Set Membership: |
|

znf106a
Genomic Map