Assay Details
| Gene Symbol: znf106a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003200378.4 | XP_003200426.2 | 9 - 10 | 2934 | 74 | |
| GenBank mRNA | BC076401.1 | - | 9 - 10 | 2932 | 74 | |
More Information
Set Membership: |
|

znf106a
Genomic Map