Assay Details
Gene Symbol: c8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003496.1 | NP_001003496.1 | 10 - 11 | 1621 | 54 | |
XM_009296648.3 | XP_009294923.1 | 10 - 11 | 1691 | 54 | ||
GenBank mRNA | BC078409.1 | - | 10 - 11 | 1621 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
