Assay Details
| Gene Symbol: cdk5r2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003508.1 | NP_001003508.1 | 1 | 1051 | 104 | |
| GenBank mRNA | BC077155.1 | - | 1 | 1051 | 104 | |
| BC164746.1 | - | 1 | 865 | 104 | ||
More Information
Set Membership: |
|

cdk5r2b
Genomic Map