Assay Details
| Gene Symbol: rab35b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003548.2 | NP_001003548.1 | 1 - 2 | 222 | 79 | |
| GenBank mRNA | BC077124.1 | - | 1 - 2 | 226 | 79 | |
More Information
Set Membership: |
|

rab35b
Genomic Map