Assay Details
| Gene Symbol: kptn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003642.2 | NP_001003642.1 | 7 - 8 | 766 | 58 | |
| GenBank mRNA | BC078206.1 | - | 7 - 8 | 770 | 58 | |
More Information
Set Membership: |
|

kptn
Genomic Map