Assay Details
Gene Symbol: cd81b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003735.1 | NP_001003735.1 | 1 - 2 | 90 | 92 | |
GenBank mRNA | BC078389.1 | - | 1 - 2 | 90 | 92 | |
BC165081.1 | - | 1 - 2 | 99 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
