Assay Details
| Gene Symbol: cd81b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003735.1 | NP_001003735.1 | 2 - 3 | 205 | 84 | |
| GenBank mRNA | BC078389.1 | - | 2 - 3 | 205 | 84 | |
| BC165081.1 | - | 2 - 3 | 214 | 84 | ||
More Information
Set Membership: |
|

cd81b
Genomic Map