Assay Details
Gene Symbol: cd81b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003735.1 | NP_001003735.1 | 3 - 4 | 294 | 74 | |
GenBank mRNA | BC078389.1 | - | 3 - 4 | 294 | 74 | |
BC165081.1 | - | 3 - 4 | 303 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
