Assay Details
Gene Symbol: cd81b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003735.1 | NP_001003735.1 | 4 - 5 | 378 | 124 | |
GenBank mRNA | BC078389.1 | - | 4 - 5 | 378 | 124 | |
BC165081.1 | - | 4 - 5 | 387 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
