Assay Details
| Gene Symbol: glmp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004580.1 | NP_001004580.1 | 5 - 6 | 1148 | 75 | |
| GenBank mRNA | BC081645.1 | - | 5 - 6 | 1148 | 75 | |
More Information
Set Membership: |
|

glmp
Genomic Map