Assay Details
| Gene Symbol: polr3gla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004639.1 | NP_001004639.2 | 2 - 3 | 235 | 113 | |
| NM_001244802.1 | NP_001231731.1 | 2 - 3 | 263 | 113 | ||
| GenBank mRNA | BC081380.1 | - | 2 - 3 | 235 | 113 | |
| DV591542.1 | - | 2 - 3 | 280 | 113 | ||
More Information
Set Membership: |
|

polr3gla
Genomic Map