Assay Details
| Gene Symbol: tcf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005399.1 | NP_001005399.1 | 5 - 6 | 550 | 64 | |
| XM_005159180.4 | XP_005159237.1 | 5 - 6 | 656 | 64 | ||
| GenBank mRNA | BC080235.1 | - | 5 - 6 | 550 | 64 | |
More Information
Set Membership: |
|

tcf25
Genomic Map