Assay Details
| Gene Symbol: amt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006021.1 | NP_001006021.1 | 7 - 8 | 1008 | 70 | |
| GenBank mRNA | BC083400.1 | - | 7 - 8 | 1008 | 70 | |
| BC164008.1 | - | 7 - 8 | 932 | 70 | ||
| DQ104411.1 | - | 7 - 8 | 1001 | 70 | ||
More Information
Set Membership: |
|

amt
Genomic Map