Assay Details
| Gene Symbol: polr1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007153.1 | NP_001007154.1 | 1 - 2 | 270 | 73 | |
| XM_017352209.2 | XP_017207698.1 | 1 - 2 | 350 | 73 | ||
| GenBank mRNA | AY648783.1 | - | 1 - 2 | 270 | 73 | |
| BC064328.1 | - | 1 - 2 | 257 | 73 | ||
More Information
Set Membership: |
|

polr1f
Genomic Map