Assay Details
| Gene Symbol: polr1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007153.1 | NP_001007154.1 | 3 - 4 | 626 | 134 | |
| XM_017352209.2 | XP_017207698.1 | 3 - 4 | 706 | 134 | ||
| GenBank mRNA | AY648783.1 | - | 3 - 4 | 626 | 134 | |
| BC056557.1 | - | 3 - 4 | 617 | 134 | ||
| BC064328.1 | - | 3 - 4 | 613 | 134 | ||
More Information
Set Membership: |
|

polr1f
Genomic Map