Assay Details
| Gene Symbol: gsto2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007372.1 | NP_001007373.1 | 6 - 7 | 630 | 64 | |
| GenBank mRNA | BC085467.1 | - | 6 - 7 | 630 | 64 | |
| BC165433.1 | - | 5 - 6 | 605 | 64 | ||
More Information
Set Membership: |
|

gsto2
Genomic Map