Assay Details
| Gene Symbol: cdc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007409.1 | NP_001007410.1 | 3 - 4 | 368 | 80 | |
| XM_005171160.4 | XP_005171217.1 | 3 - 4 | 513 | 80 | ||
| XM_005171161.4 | XP_005171218.1 | 3 - 4 | 513 | 80 | ||
| XM_005171162.4 | XP_005171219.1 | 3 - 4 | 513 | 80 | ||
| GenBank mRNA | BC085380.1 | - | 3 - 4 | 368 | 80 | |
More Information
Set Membership: |
|

cdc7
Genomic Map