Assay Details
| Gene Symbol: fgf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007761.1 | NP_001007762.1 | 2 - 3 | 326 | 90 | |
| XM_009293389.3 | XP_009291664.1 | 3 - 4 | 587 | 90 | ||
| GenBank mRNA | AB194727.1 | - | 2 - 3 | 326 | 90 | |
| BC162865.1 | - | 2 - 3 | 400 | 90 | ||
| BC162898.1 | - | 2 - 3 | 400 | 90 | ||
More Information
Set Membership: |
|

fgf7
Genomic Map