Assay Details
| Gene Symbol: rmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001362546.1 | NP_001349475.1 | 16 - 17 | 128 | ||
| GenBank mRNA | BC086740.1 | - | 15 - 16 | 1667 | 128 | |
More Information
Set Membership: |
|

rmc1
Genomic Map