Assay Details
| Gene Symbol: mnx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009885.2 | NP_001009885.1 | 1 - 2 | 627 | 73 | |
| GenBank mRNA | AY445044.1 | - | 1 - 2 | 495 | 73 | |
| BC093243.1 | - | 1 - 2 | 627 | 73 | ||
More Information
Set Membership: |
|

mnx1
Genomic Map