Assay Details
| Gene Symbol: wars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013323.2 | NP_001013341.1 | 2 - 3 | 372 | 56 | |
| GenBank mRNA | BC090784.1 | - | 2 - 3 | 376 | 56 | |
| BC164194.1 | - | 2 - 3 | 394 | 56 | ||
More Information
Set Membership: |
|

wars2
Genomic Map