Assay Details
| Gene Symbol: rpp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013479.1 | NP_001013497.1 | 5 - 6 | 412 | 124 | |
| XM_021480064.1 | XP_021335739.1 | 5 - 6 | 265 | 124 | ||
| GenBank mRNA | BC091449.1 | - | 5 - 6 | 412 | 124 | |
| BC164838.1 | - | 5 - 6 | 362 | 124 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021480064.1 transcripts.
Set Membership: |
|

rpp30
Genomic Map