Assay Details
| Gene Symbol: eif3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017583.1 | NP_001017583.1 | 4 - 5 | 451 | 67 | |
| XM_017350953.2 | XP_017206442.1 | 4 - 5 | 800 | 67 | ||
| GenBank mRNA | BC093002.1 | - | 4 - 5 | 451 | 67 | |
More Information
Set Membership: |
|

eif3k
Genomic Map