Assay Details
| Gene Symbol: stoml3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017825.1 | NP_001017825.1 | 5 - 6 | 556 | 88 | |
| GenBank mRNA | BC092792.1 | - | 5 - 6 | 556 | 88 | |
More Information
Set Membership: |
|

stoml3b
Genomic Map