Assay Details
| Gene Symbol: ankrd52a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018154.1 | NP_001018164.1 | 10 - 11 | 1184 | 95 | |
| GenBank mRNA | BC095664.1 | - | 10 - 11 | 1184 | 95 | |
| BC165234.1 | - | 10 - 11 | 1131 | 95 | ||
More Information
Set Membership: |
|

ankrd52a
Genomic Map