Assay Details
| Gene Symbol: impa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020572.2 | NP_001018408.1 | 3 - 4 | 417 | 66 | |
| XM_021470172.1 | XP_021325847.1 | 3 - 4 | 440 | 66 | ||
| GenBank mRNA | BC095198.1 | - | 3 - 4 | 417 | 66 | |
| BC165539.1 | - | 3 - 4 | 339 | 66 | ||
More Information
Set Membership: |
|

impa2
Genomic Map