Assay Details
| Gene Symbol: pip5k1aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020602.1 | NP_001018438.1 | 13 - 14 | 1744 | 80 | |
| GenBank mRNA | BC095318.1 | - | 13 - 14 | 1744 | 80 | |
More Information
Set Membership: |
|

pip5k1aa
Genomic Map