Assay Details
| Gene Symbol: parp12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115086.1 | NP_001108558.1 | 1 - 2 | 446 | 81 | |
| GenBank mRNA | BC095326.1 | - | 1 - 2 | 416 | 81 | |
| BC165181.1 | - | 1 - 2 | 359 | 81 | ||
More Information
Set Membership: |
|

parp12b
Genomic Map