Assay Details
| Gene Symbol: parp12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115086.1 | NP_001108558.1 | 7 - 8 | 1345 | 152 | |
| XM_009300122.3 | XP_009298397.1 | 6 - 7 | 955 | 152 | ||
| GenBank mRNA | BC095326.1 | - | 7 - 8 | 1315 | 152 | |
| BC165181.1 | - | 7 - 8 | 1258 | 152 | ||
More Information
Set Membership: |
|

parp12b
Genomic Map