Assay Details
| Gene Symbol: pudp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020615.1 | NP_001018451.1 | 2 - 3 | 309 | 102 | |
| GenBank mRNA | BC095345.1 | - | 2 - 3 | 309 | 102 | |
| BC165028.1 | - | 2 - 3 | 279 | 102 | ||
More Information
Set Membership: |
|

pudp
Genomic Map