Assay Details
| Gene Symbol: ddx47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020686.1 | NP_001018522.1 | 5 - 6 | 933 | 91 | |
| GenBank mRNA | BC095776.1 | - | 5 - 6 | 933 | 91 | |
More Information
Set Membership: |
|

ddx47
Genomic Map