Assay Details
| Gene Symbol: fbxo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346202.1 | NP_001333131.1 | 5 - 6 | 686 | 65 | |
| NR_144396.1 | 5 - 6 | 686 | 65 | |||
| GenBank mRNA | BC095831.1 | - | 5 - 6 | 681 | 65 | |
| CN501448.1 | - | 5 - 6 | 659 | 65 | ||
More Information
Set Membership: |
|

fbxo3
Genomic Map