Assay Details
| Gene Symbol: il10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020785.2 | NP_001018621.2 | 3 - 4 | 391 | 70 | |
| GenBank mRNA | BC163031.1 | - | 3 - 4 | 481 | 70 | |
| BC163038.1 | - | 3 - 4 | 481 | 70 | ||
More Information
Set Membership: |
|

il10
Genomic Map